Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTGCCCCATATCCTAATACCAG[C/T]GAATCTCAGGCTGGTTATAGGGTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300457 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NHS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NHS - NHS actin remodeling regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136024.3 | Intron | NP_001129496.1 | ||||
NM_001291867.1 | Intron | NP_001278796.1 | ||||
NM_001291868.1 | Intron | NP_001278797.1 | ||||
NM_198270.3 | Intron | NP_938011.1 | ||||
XM_011545528.2 | Intron | XP_011543830.1 |