Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGGGACATGTCCCATTTTCTTCT[A/G]TATTCAGGAAGAGTCTCTGTAGAAT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 612477 MIM: 609380 | |||||||||||||||||||||||
Literature Links: |
IQCG PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
IQCG - IQ motif containing G | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LMLN - leishmanolysin like peptidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136049.2 | Intron | NP_001129521.2 | ||||
NM_033029.3 | Intron | NP_149018.2 | ||||
XM_011513270.1 | Intron | XP_011511572.1 | ||||
XM_011513271.1 | Intron | XP_011511573.1 | ||||
XM_017007428.1 | Intron | XP_016862917.1 |