Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCATGGAAAACATGTGCATATTA[A/G]AACTTTAACATTTAAATTACAGTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601081 | ||||||||||||||||||||
Literature Links: |
ABCD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABCD2 - ATP binding cassette subfamily D member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005164.3 | 4928 | UTR 3 | NP_005155.1 | |||
XM_011538027.2 | 4928 | UTR 3 | XP_011536329.1 | |||
XM_017018992.1 | 4928 | Intron | XP_016874481.1 | |||
XM_017018993.1 | 4928 | Intron | XP_016874482.1 | |||
XM_017018994.1 | 4928 | Intron | XP_016874483.1 |