Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGTCCTGAGCCCCAGTAATGCAA[C/T]AGCTCCTCATTTTCTCCTTCCCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102578 MIM: 608326 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PML PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PML - promyelocytic leukemia | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002675.3 | Intron | NP_002666.1 | ||||
NM_033238.2 | Intron | NP_150241.2 | ||||
NM_033239.2 | Intron | NP_150242.1 | ||||
NM_033240.2 | Intron | NP_150243.2 | ||||
NM_033244.3 | Intron | NP_150247.2 | ||||
NM_033246.2 | Intron | NP_150249.1 | ||||
NM_033247.2 | Intron | NP_150250.2 | ||||
NM_033249.2 | Intron | NP_150252.1 | ||||
NM_033250.2 | Intron | NP_150253.2 |
STOML1 - stomatin like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |