Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTCATTGCACAAAGTATTGTTTA[G/T]AAGGTGCTTTTAGGAGTATCTTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615876 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RSPH3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RSPH3 - radial spoke 3 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031924.4 | Intron | NP_114130.3 | ||||
XM_005267153.4 | Intron | XP_005267210.1 | ||||
XM_017011347.1 | Intron | XP_016866836.1 |
Set Membership: |
HapMap |