Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAAAATGTAATTTAAAAAATGCA[A/G]TGAGTCTAGACAACATGGTGAGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF149 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RNF149 - ring finger protein 149 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173647.3 | 1984 | Intron | NP_775918.2 | |||
XM_005263920.2 | 1984 | UTR 3 | XP_005263977.1 | |||
XM_005263921.3 | 1984 | Intron | XP_005263978.1 | |||
XM_011510990.2 | 1984 | UTR 3 | XP_011509292.1 |
SNORD89 - small nucleolar RNA, C/D box 89 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |