Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAAGTTAGAGATAAATATAGGGG[A/T]TAAGTGAGAAATTCTCCCTGGGGCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601081 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCD2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ABCD2 - ATP binding cassette subfamily D member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005164.3 | Intron | NP_005155.1 | ||||
XM_011538027.2 | Intron | XP_011536329.1 | ||||
XM_017018992.1 | Intron | XP_016874481.1 | ||||
XM_017018993.1 | Intron | XP_016874482.1 | ||||
XM_017018994.1 | Intron | XP_016874483.1 |
Set Membership: |
HapMap |