Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
57 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609473 MIM: 600087 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CGN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CGN - cingulin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR554 - microRNA 554 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TUFT1 - tuftelin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126337.1 | Intron | NP_001119809.1 | ||||
NM_001301317.1 | Intron | NP_001288246.1 | ||||
NM_020127.2 | Intron | NP_064512.1 | ||||
XM_011509961.2 | Intron | XP_011508263.1 | ||||
XM_017002223.1 | Intron | XP_016857712.1 | ||||
XM_017002224.1 | Intron | XP_016857713.1 |
Set Membership: |
HapMap |