Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612793 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTDSS2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
PTDSS2 - phosphatidylserine synthase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005253153.1 | 2737 | Intron | XP_005253210.1 | |||
XM_011520391.2 | 2737 | Intron | XP_011518693.1 | |||
XM_011520392.2 | 2737 | Intron | XP_011518694.1 | |||
XM_011520393.2 | 2737 | Intron | XP_011518695.1 | |||
XM_017018368.1 | 2737 | UTR 5 | XP_016873857.1 | |||
XM_017018369.1 | 2737 | Intron | XP_016873858.1 | |||
XM_017018370.1 | 2737 | Intron | XP_016873859.1 | |||
XM_017018371.1 | 2737 | Intron | XP_016873860.1 |
Set Membership: |
HapMap |