Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTAGACTATAATACAAATGGAGTC[G/T]GAGTCTGAAAACAACTCATATCTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601769 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VDR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VDR - vitamin D (1,25- dihydroxyvitamin D3) receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000376.2 | 3232 | UTR 3 | NP_000367.1 | |||
NM_001017535.1 | 3232 | UTR 3 | NP_001017535.1 | |||
NM_001017536.1 | 3232 | UTR 3 | NP_001017536.1 | |||
XM_006719587.3 | 3232 | Intron | XP_006719650.1 | |||
XM_011538720.2 | 3232 | Intron | XP_011537022.1 |