Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATGAGGGTAGCCCGTACTGGGTG[A/C]CGACGCTGGCAGCGGAGAGACAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614964 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELFN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ELFN1 - extracellular leucine rich repeat and fibronectin type III domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128636.2 | Intron | NP_001122108.1 | ||||
XM_006715725.3 | Intron | XP_006715788.1 | ||||
XM_006715726.2 | Intron | XP_006715789.1 | ||||
XM_006715727.3 | Intron | XP_006715790.1 | ||||
XM_011515397.2 | Intron | XP_011513699.1 | ||||
XM_011515398.2 | Intron | XP_011513700.1 | ||||
XM_011515401.2 | Intron | XP_011513703.1 | ||||
XM_017012203.1 | Intron | XP_016867692.1 |
Set Membership: |
HapMap |