Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACACATGTTGAGGGAAAATTTTTA[C/T]TGATTCACAAAAAGGAAGACAGTTT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
|||||||||||||||||||||||
Phenotype: |
MIM: 614698 MIM: 602869 | |||||||||||||||||||||||
Literature Links: |
COX20 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
COX20 - COX20, cytochrome c oxidase assembly factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001312871.1 | 1819 | UTR 3 | NP_001299800.1 | |||
NM_001312872.1 | 1819 | UTR 3 | NP_001299801.1 | |||
NM_001312873.1 | 1819 | UTR 3 | NP_001299802.1 | |||
NM_001312874.1 | 1819 | UTR 3 | NP_001299803.1 | |||
NM_198076.5 | 1819 | UTR 3 | NP_932342.1 |
HNRNPU - heterogeneous nuclear ribonucleoprotein U | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004501.3 | 1819 | Intron | NP_004492.2 | |||
NM_031844.2 | 1819 | Intron | NP_114032.2 | |||
XM_017001115.1 | 1819 | UTR 3 | XP_016856604.1 | |||
XM_017001116.1 | 1819 | UTR 3 | XP_016856605.1 | |||
XM_017001117.1 | 1819 | UTR 3 | XP_016856606.1 |
SNORA100 - small nucleolar RNA, H/ACA box 100 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |