Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCTCATTGTTCATGATGCACAGG[A/C]TGGGTTACAGGGAAGGAGGTGAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 104221 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADRA1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADRA1A - adrenoceptor alpha 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000680.3 | 13074 | Intron | NP_000671.2 | |||
NM_001322502.1 | 13074 | Intron | NP_001309431.1 | |||
NM_001322503.1 | 13074 | Intron | NP_001309432.1 | |||
NM_001322504.1 | 13074 | Intron | NP_001309433.1 | |||
NM_033302.3 | 13074 | Intron | NP_150645.2 | |||
NM_033303.4 | 13074 | Intron | NP_150646.3 | |||
NM_033304.3 | 13074 | Intron | NP_150647.2 | |||
XM_006716292.3 | 13074 | UTR 3 | XP_006716355.1 | |||
XM_006716293.3 | 13074 | Intron | XP_006716356.1 | |||
XM_011544411.2 | 13074 | Intron | XP_011542713.1 | |||
XM_011544412.2 | 13074 | Intron | XP_011542714.1 | |||
XM_017013094.1 | 13074 | UTR 3 | XP_016868583.1 | |||
XM_017013095.1 | 13074 | UTR 3 | XP_016868584.1 | |||
XM_017013096.1 | 13074 | Intron | XP_016868585.1 |