Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAACCTCATCCTGTGGTCTGATG[C/T]CTACAGGGGTTGGAAGATAGTTTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
45 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179510 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB3B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAB3B - RAB3B, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002867.3 | 10600 | UTR 3 | NP_002858.2 | |||
XM_017001958.1 | 10600 | Intron | XP_016857447.1 |