Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATGCCTCACCTTGGAGACCTATT[C/T]TAATGAATGTCAATTAGTGAATGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605460 MIM: 607544 | ||||||||||||||||||||
Literature Links: |
ABCG8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABCG8 - ATP binding cassette subfamily G member 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRPPRC - leucine rich pentatricopeptide repeat containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133259.3 | 6089 | UTR 3 | NP_573566.2 | |||
XM_006711915.2 | 6089 | Intron | XP_006711978.1 | |||
XM_006711916.3 | 6089 | Intron | XP_006711979.1 | |||
XM_011532473.1 | 6089 | Intron | XP_011530775.1 | |||
XM_017003117.1 | 6089 | Intron | XP_016858606.1 |