Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAAGAAATTCATTAAATGTTGTA[C/G]AGATAGAGCCAAGCAGCGCTCCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608399 | ||||||||||||||||||||
Literature Links: |
CSMD3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CSMD3 - CUB and Sushi multiple domains 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052900.2 | 13175 | UTR 3 | NP_443132.3 | |||
NM_198123.1 | 13175 | UTR 3 | NP_937756.1 | |||
NM_198124.1 | 13175 | UTR 3 | NP_937757.1 | |||
XM_011516815.2 | 13175 | UTR 3 | XP_011515117.1 | |||
XM_011516816.2 | 13175 | UTR 3 | XP_011515118.1 | |||
XM_017013008.1 | 13175 | UTR 3 | XP_016868497.1 | |||
XM_017013009.1 | 13175 | UTR 3 | XP_016868498.1 | |||
XM_017013010.1 | 13175 | UTR 3 | XP_016868499.1 | |||
XM_017013011.1 | 13175 | UTR 3 | XP_016868500.1 | |||
XM_017013012.1 | 13175 | UTR 3 | XP_016868501.1 |