Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606453 MIM: 604357 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC729558 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC729558 - uncharacterized LOC729558 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRBA - LPS responsive beige-like anchor protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199282.2 | 181 | Intron | NP_001186211.2 | |||
NM_006726.4 | 181 | Intron | NP_006717.2 | |||
XM_005263372.3 | 181 | Intron | XP_005263429.1 | |||
XM_005263373.2 | 181 | Intron | XP_005263430.1 | |||
XM_005263374.3 | 181 | Intron | XP_005263431.1 | |||
XM_005263375.3 | 181 | Intron | XP_005263432.1 | |||
XM_011532434.2 | 181 | Intron | XP_011530736.1 | |||
XM_017008872.1 | 181 | Intron | XP_016864361.1 | |||
XM_017008873.1 | 181 | Intron | XP_016864362.1 | |||
XM_017008874.1 | 181 | Intron | XP_016864363.1 |
MAB21L2 - mab-21 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006439.4 | 181 | UTR 5 | NP_006430.1 |