Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCTATCCATGGAGGATCCTCTTA[C/T]CCTCCACATCTCATCTATCCTCAGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613833 MIM: 180480 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KANSL1L PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KANSL1L - KAT8 regulatory NSL complex subunit 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPE - ribulose-5-phosphate-3-epimerase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278282.1 | Intron | NP_001265211.1 | ||||
NM_001278283.1 | Intron | NP_001265212.1 | ||||
NM_001278285.1 | Intron | NP_001265214.1 | ||||
NM_001278286.1 | Intron | NP_001265215.1 | ||||
NM_001278288.1 | Intron | NP_001265217.1 | ||||
NM_001278289.1 | Intron | NP_001265218.1 | ||||
NM_001318926.1 | Intron | NP_001305855.1 | ||||
NM_001318927.1 | Intron | NP_001305856.1 | ||||
NM_001318928.1 | Intron | NP_001305857.1 | ||||
NM_001318929.1 | Intron | NP_001305858.1 | ||||
NM_001318930.1 | Intron | NP_001305859.1 | ||||
NM_001318931.1 | Intron | NP_001305860.1 | ||||
NM_006916.2 | Intron | NP_008847.1 | ||||
NM_199229.2 | Intron | NP_954699.1 | ||||
XM_006712677.3 | Intron | XP_006712740.1 |