Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCACAAGAAAGGAGAGAGGATAAA[A/G]ACTGAAGTGTGTGCCAAAGTCATTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
32 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612666 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DSTYK PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DSTYK - dual serine/threonine and tyrosine protein kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015375.2 | 7621 | UTR 3 | NP_056190.1 | |||
NM_199462.2 | 7621 | UTR 3 | NP_955749.1 | |||
XM_011509392.2 | 7621 | UTR 3 | XP_011507694.1 | |||
XM_011509393.2 | 7621 | UTR 3 | XP_011507695.1 | |||
XM_011509394.2 | 7621 | UTR 3 | XP_011507696.1 |
Set Membership: |
HapMap |