Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTAGATAAATAAAGATCCATAT[A/C]CAGTGTTGGTATTTTGTAGAAAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603654 | ||||||||||||||||||||
Literature Links: |
SLC16A7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC16A7 - solute carrier family 16 member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270622.1 | Intron | NP_001257551.1 | ||||
NM_001270623.1 | Intron | NP_001257552.1 | ||||
NM_004731.4 | Intron | NP_004722.2 | ||||
XM_005269231.4 | Intron | XP_005269288.1 | ||||
XM_011538989.2 | Intron | XP_011537291.1 | ||||
XM_011538990.2 | Intron | XP_011537292.1 | ||||
XM_011538991.2 | Intron | XP_011537293.1 | ||||
XM_011538992.2 | Intron | XP_011537294.1 | ||||
XM_011538993.2 | Intron | XP_011537295.1 | ||||
XM_011538995.2 | Intron | XP_011537297.1 | ||||
XM_017020224.1 | Intron | XP_016875713.1 | ||||
XM_017020225.1 | Intron | XP_016875714.1 | ||||
XM_017020226.1 | Intron | XP_016875715.1 | ||||
XM_017020227.1 | Intron | XP_016875716.1 |