Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTCTTCTTTCCTGCCTTCCATT[C/T]CCAAAATTGTTCTGGAAGGGCCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607002 | ||||||||||||||||||||
Literature Links: |
PROK2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PROK2 - prokineticin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126128.1 | 322 | Silent Mutation | GGA,GGG | G,G 78 | NP_001119600.1 | |
NM_021935.3 | 322 | Intron | NP_068754.1 | |||
XM_017006974.1 | 322 | Silent Mutation | GGA,GGG | G,G 58 | XP_016862463.1 |