Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCATGGCCGAGGCAGCGCTAGTGA[A/T]TACGCCGCAGGTGAGAGCGGAGTCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF549 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF549 - zinc finger protein 549 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199295.1 | 276 | Missense Mutation | AAT,ATT | N,I 8 | NP_001186224.1 | |
NM_153263.2 | 276 | Missense Mutation | AAT,ATT | N,I 8 | NP_694995.2 | |
XM_006723128.3 | 276 | UTR 5 | XP_006723191.1 |