Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAGTCTTCATCCTCTCTACGATA[C/T]TCTTCCTTTGAATGTCCTTTGGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF417 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF417 - zinc finger protein 417 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297734.1 | 191 | Intron | NP_001284663.1 | |||
NM_152475.2 | 191 | Intron | NP_689688.2 | |||
XM_011526473.2 | 191 | UTR 5 | XP_011524775.1 | |||
XM_017026315.1 | 191 | Intron | XP_016881804.1 |
ZNF418 - zinc finger protein 418 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |