Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGCTTACATAAACTACAATAGAA[A/C]CAGATGTTTGCTGGTTGTCTAGTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604762 | ||||||||||||||||||||
Literature Links: |
SWAP70 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SWAP70 - SWAP switching B-cell complex 70kDa subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297714.1 | Intron | NP_001284643.1 | ||||
NM_015055.3 | Intron | NP_055870.2 | ||||
XM_005252830.4 | Intron | XP_005252887.1 |