Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAGCGTCCATAGGGTTTTATGTT[G/T]TTCATGTTTGAAATCCTGTTGTCAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
25 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603185 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NASP PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NASP - nuclear autoantigenic sperm protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195193.1 | Intron | NP_001182122.1 | ||||
NM_002482.3 | Intron | NP_002473.2 | ||||
NM_152298.3 | Intron | NP_689511.2 | ||||
XM_005270888.2 | Intron | XP_005270945.1 | ||||
XM_005270889.2 | Intron | XP_005270946.1 | ||||
XM_011541509.1 | Intron | XP_011539811.1 | ||||
XM_017001356.1 | Intron | XP_016856845.1 |
Set Membership: |
HapMap |