Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTCTGCTTTATGTCTAATAAGTA[C/T]TGTGGGAAATAGAAAAGTAGGTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100506747 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC100506747 - alpha-1,3-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase-like protein LOC641515 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM183A - transmembrane protein 183A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322955.1 | Intron | NP_001309884.1 | ||||
NM_001322956.1 | Intron | NP_001309885.1 | ||||
NM_001322957.1 | Intron | NP_001309886.1 | ||||
NM_001322958.1 | Intron | NP_001309887.1 | ||||
NM_001322959.1 | Intron | NP_001309888.1 | ||||
NM_138391.4 | Intron | NP_612400.3 |