Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTAACCTCATATATGAAAATACTG[C/G]ACCCACTGCTTCTGAGTTAGATACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300747 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
STS - steroid sulfatase (microsomal), isozyme S | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000351.5 | Intron | NP_000342.2 | ||||
NM_001320750.1 | Intron | NP_001307679.1 | ||||
NM_001320751.1 | Intron | NP_001307680.1 | ||||
NM_001320752.1 | Intron | NP_001307681.1 | ||||
NM_001320753.1 | Intron | NP_001307682.1 | ||||
NM_001320754.1 | Intron | NP_001307683.1 |
Set Membership: |
HapMap |