Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGATAAATTGCTCAAGGACCTGGA[C/T]GCCAATGGAGATGCCCAGGTGGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600614 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC339988 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC339988 - uncharacterized LOC339988 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
S100P - S100 calcium binding protein P | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005980.2 | 280 | Silent Mutation | GAC,GAT | D,D 62 | NP_005971.1 |