Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCACATATTCTCTGGGGGCATA[C/G]AGGGGTACAGAAACATTCTCATGAT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 606775 | |||||||||||||||||||||||
Literature Links: |
CSPG5 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CSPG5 - chondroitin sulfate proteoglycan 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206942.1 | Intron | NP_001193871.1 | ||||
NM_001206943.1 | Intron | NP_001193872.1 | ||||
NM_001206944.1 | Intron | NP_001193873.1 | ||||
NM_001206945.1 | Intron | NP_001193874.1 | ||||
NM_006574.3 | Intron | NP_006565.2 |