Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTTAGCTGCTAGGGTGACCTTTC[C/T]CCTCCTTAAAGCCTCCCATGGGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609748 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBL7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
UBL7 - ubiquitin like 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286739.1 | Intron | NP_001273668.1 | ||||
NM_001286740.1 | Intron | NP_001273669.1 | ||||
NM_001286741.1 | Intron | NP_001273670.1 | ||||
NM_001286742.1 | Intron | NP_001273671.1 | ||||
NM_032907.4 | Intron | NP_116296.1 | ||||
NM_201265.1 | Intron | NP_957717.1 | ||||
XM_011522124.2 | Intron | XP_011520426.1 |