Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAGAGGAAAAACGGAAGAGGGAA[A/T]GTTGGGGAAAGCTTCCTAGAGGGCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116900 MIM: 610595 MIM: 600560 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CKS1B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CKS1B - CDC28 protein kinase regulatory subunit 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001826.2 | Intron | NP_001817.1 |
FLAD1 - flavin adenine dinucleotide synthetase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4258 - microRNA 4258 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SHC1 - SHC adaptor protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |