Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCATGTCCTTCCTAAATCTCTCT[C/T]ATAAGCAGCTATGGCTTTATAAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613439 MIM: 607055 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNST PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CNST - consortin, connexin sorting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001139459.1 | Intron | NP_001132931.1 | ||||
NM_152609.2 | Intron | NP_689822.2 | ||||
XM_005273081.3 | Intron | XP_005273138.2 | ||||
XM_005273083.4 | Intron | XP_005273140.1 | ||||
XM_011544110.2 | Intron | XP_011542412.1 | ||||
XM_011544111.1 | Intron | XP_011542413.1 | ||||
XM_011544112.1 | Intron | XP_011542414.1 | ||||
XM_011544113.1 | Intron | XP_011542415.1 | ||||
XM_011544114.2 | Intron | XP_011542416.1 | ||||
XM_017000487.1 | Intron | XP_016855976.1 | ||||
XM_017000488.1 | Intron | XP_016855977.1 | ||||
XM_017000489.1 | Intron | XP_016855978.1 |
TFB2M - transcription factor B2, mitochondrial | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |