Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATATATAAATATATTCTCAAAGC[A/T]AAACTGTAAACGCTACAGATAAAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 153243 | ||||||||||||||||||||
Literature Links: |
TNFRSF8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TNFRSF8 - TNF receptor superfamily member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243.4 | Intron | NP_001234.3 | ||||
NM_001281430.2 | Intron | NP_001268359.2 | ||||
XM_011542441.2 | Intron | XP_011540743.1 | ||||
XM_011542443.2 | Intron | XP_011540745.1 |