Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608541 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP32 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARHGAP32 - Rho GTPase activating protein 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142685.1 | 6512 | UTR 3 | NP_001136157.1 | |||
NM_014715.3 | 6512 | UTR 3 | NP_055530.2 | |||
XM_005271736.4 | 6512 | UTR 3 | XP_005271793.2 | |||
XM_011543072.2 | 6512 | UTR 3 | XP_011541374.1 | |||
XM_011543073.2 | 6512 | UTR 3 | XP_011541375.2 | |||
XM_011543075.2 | 6512 | UTR 3 | XP_011541377.1 | |||
XM_017018595.1 | 6512 | UTR 3 | XP_016874084.1 | |||
XM_017018596.1 | 6512 | UTR 3 | XP_016874085.1 | |||
XM_017018597.1 | 6512 | Intron | XP_016874086.1 |
Set Membership: |
HapMap |