Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603862 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCNT2 - cyclin T2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001241.3 | Intron | NP_001232.1 | ||||
NM_001320748.1 | Intron | NP_001307677.1 | ||||
NM_001320749.1 | Intron | NP_001307678.1 | ||||
NM_058241.2 | Intron | NP_490595.1 | ||||
XM_017005227.1 | Intron | XP_016860716.1 | ||||
XM_017005228.1 | Intron | XP_016860717.1 | ||||
XM_017005229.1 | Intron | XP_016860718.1 | ||||
XM_017005230.1 | Intron | XP_016860719.1 | ||||
XM_017005231.1 | Intron | XP_016860720.1 | ||||
XM_017005232.1 | Intron | XP_016860721.1 | ||||
XM_017005233.1 | Intron | XP_016860722.1 |
CCNT2-AS1 - CCNT2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |