Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGACTTGGTCCCATTGTACAAACT[G/T]GGAAACTGAGGCTTGGAGGAACACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613368 MIM: 610831 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CARNS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CARNS1 - carnosine synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166222.1 | Intron | NP_001159694.1 | ||||
NM_020811.1 | Intron | NP_065862.1 | ||||
XM_011545189.2 | Intron | XP_011543491.1 | ||||
XM_011545191.2 | Intron | XP_011543493.1 | ||||
XM_017018054.1 | Intron | XP_016873543.1 | ||||
XM_017018055.1 | Intron | XP_016873544.1 |
TBC1D10C - TBC1 domain family member 10C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |