Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCAGTGCTGGCTCTTGGCCCTG[A/T]CACTGTGTCTGTTTTATCCTGGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OGDHL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OGDHL - oxoglutarate dehydrogenase-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143996.1 | 3189 | UTR 3 | NP_001137468.1 | |||
NM_001143997.1 | 3189 | UTR 3 | NP_001137469.1 | |||
NM_018245.2 | 3189 | UTR 3 | NP_060715.2 | |||
XM_011539946.1 | 3189 | UTR 3 | XP_011538248.1 | |||
XM_011539947.1 | 3189 | UTR 3 | XP_011538249.1 | |||
XM_017016401.1 | 3189 | UTR 3 | XP_016871890.1 | |||
XM_017016402.1 | 3189 | UTR 3 | XP_016871891.1 | |||
XM_017016403.1 | 3189 | UTR 3 | XP_016871892.1 |