Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTCTCTTTCGCCACCAAGATCC[C/T]GGGGATCCAGGACCTGGAATCAGAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615920 MIM: 613175 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRR11 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PRR11 - proline rich 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMG8 - SMG8, nonsense mediated mRNA decay factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018149.6 | 297 | Silent Mutation | CCC,CCT | P,P 85 | NP_060619.4 |