Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAGAGCACAAATGAACAAAACTG[C/T]CAATTTGTTAATCCAAAAGAATTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601584 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTP4A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTP4A2 - protein tyrosine phosphatase type IVA, member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195100.1 | Intron | NP_001182029.1 | ||||
NM_001195101.1 | Intron | NP_001182030.1 | ||||
NM_080391.3 | Intron | NP_536316.1 | ||||
XM_005271231.3 | Intron | XP_005271288.1 | ||||
XM_005271232.3 | Intron | XP_005271289.1 | ||||
XM_017002401.1 | Intron | XP_016857890.1 |