Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCCCTATGTTGCCCAGTGGGCA[G/T]GGGGTATCCTCCTCTCAGTACATGT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 611972 | |||||||||||||||||||||||
Literature Links: |
MRPS5 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
MRPS5 - mitochondrial ribosomal protein S5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF514 - zinc finger protein 514 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318005.1 | Intron | NP_001304934.1 | ||||
NM_032788.2 | Intron | NP_116177.1 | ||||
XM_006712806.2 | Intron | XP_006712869.1 |