Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGAGCACCAGCCTAGACCAGGA[C/T]GCCTCCACCTCAGCAACACCGCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610309 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBE2S PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
UBE2S - ubiquitin conjugating enzyme E2 S | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014501.2 | Intron | NP_055316.2 | ||||
XM_011526752.1 | Intron | XP_011525054.1 |
Set Membership: |
HapMap |