Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCTCTGCCCTGTACCTCACTGAA[A/T]TGTTTTAGGATTAGTGATATGGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613140 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSPAN15 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TSPAN15 - tetraspanin 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012339.3 | Intron | NP_036471.1 | ||||
XM_005269666.4 | Intron | XP_005269723.1 | ||||
XM_005269667.3 | Intron | XP_005269724.1 | ||||
XM_011539562.2 | Intron | XP_011537864.1 | ||||
XM_011539563.2 | Intron | XP_011537865.1 | ||||
XM_017016010.1 | Intron | XP_016871499.1 |