Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 151510 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITGAX PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ITGAX - integrin subunit alpha X | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000887.4 | 263 | Missense Mutation | CGG,TGG | R,W 48 | NP_000878.2 | |
NM_001286375.1 | 263 | Missense Mutation | CGG,TGG | R,W 48 | NP_001273304.1 | |
XM_011545852.1 | 263 | Missense Mutation | CGG,TGG | R,W 48 | XP_011544154.1 | |
XM_011545854.1 | 263 | Missense Mutation | CGG,TGG | R,W 48 | XP_011544156.1 |
Set Membership: |
HapMap |