Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAGGTCCGGAGATGGCTGACAGG[A/C]CAGTGACTGGGAGGAATAGGTGACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612082 | ||||||||||||||||||||
Literature Links: |
CIC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CIC - capicua transcriptional repressor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304815.1 | Intron | NP_001291744.1 | ||||
NM_015125.4 | Intron | NP_055940.3 | ||||
XM_005258673.2 | Intron | XP_005258730.1 | ||||
XM_005258674.2 | Intron | XP_005258731.1 | ||||
XM_005258675.2 | Intron | XP_005258732.1 | ||||
XM_011526660.2 | Intron | XP_011524962.1 | ||||
XM_011526661.2 | Intron | XP_011524963.1 | ||||
XM_011526662.2 | Intron | XP_011524964.1 | ||||
XM_011526663.2 | Intron | XP_011524965.1 | ||||
XM_011526664.2 | Intron | XP_011524966.1 | ||||
XM_011526665.2 | Intron | XP_011524967.1 | ||||
XM_011526666.2 | Intron | XP_011524968.1 | ||||
XM_011526667.2 | Intron | XP_011524969.1 |