Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGGGCTCTTTGCTGCAGAGCGTG[C/T]TGCAGAGCCCTGTGATGTGTCTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611379 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DIP2B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DIP2B - disco interacting protein 2 homolog B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173602.2 | Intron | NP_775873.2 | ||||
XM_005269044.1 | Intron | XP_005269101.1 | ||||
XM_006719520.3 | Intron | XP_006719583.1 | ||||
XM_006719521.3 | Intron | XP_006719584.1 | ||||
XM_011538594.2 | Intron | XP_011536896.1 | ||||
XM_011538595.2 | Intron | XP_011536897.1 | ||||
XM_011538596.2 | Intron | XP_011536898.1 | ||||
XM_017019699.1 | Intron | XP_016875188.1 | ||||
XM_017019700.1 | Intron | XP_016875189.1 | ||||
XM_017019701.1 | Intron | XP_016875190.1 | ||||
XM_017019702.1 | Intron | XP_016875191.1 | ||||
XM_017019703.1 | Intron | XP_016875192.1 | ||||
XM_017019704.1 | Intron | XP_016875193.1 | ||||
XM_017019705.1 | Intron | XP_016875194.1 |