Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGAGTGAGGTAGGTACTATCATC[A/G]CCACTTTAAGGATGAAGACAGAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
H2AFV PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
H2AFV - H2A histone family member V | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012412.4 | 2141 | Intron | NP_036544.1 | |||
NM_138635.3 | 2141 | UTR 3 | NP_619541.1 | |||
NM_201436.2 | 2141 | Intron | NP_958844.1 | |||
NM_201516.2 | 2141 | Intron | NP_958924.1 | |||
NM_201517.2 | 2141 | Intron | NP_958925.1 |
Set Membership: |
HapMap |