Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPDC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PTPDC1 - protein tyrosine phosphatase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253829.1 | 419 | Intron | NP_001240758.1 | |||
NM_001253830.1 | 419 | Intron | NP_001240759.1 | |||
NM_152422.4 | 419 | Intron | NP_689635.3 | |||
NM_177995.2 | 419 | Intron | NP_818931.1 | |||
XM_005251711.4 | 419 | UTR 5 | XP_005251768.1 | |||
XM_011518240.2 | 419 | Intron | XP_011516542.1 | |||
XM_011518241.2 | 419 | Intron | XP_011516543.1 | |||
XM_017014280.1 | 419 | Intron | XP_016869769.1 | |||
XM_017014281.1 | 419 | Intron | XP_016869770.1 | |||
XM_017014282.1 | 419 | Intron | XP_016869771.1 |
Set Membership: |
HapMap |