Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACATTCTAAGGTACTAGGGGTTA[A/G]GATGTCAACATACGAATATGGGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603404 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF169 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF169 - zinc finger protein 169 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301275.1 | 141 | Intron | NP_001288204.1 | |||
NM_003448.2 | 141 | Intron | NP_003439.2 | |||
NM_194320.3 | 141 | Intron | NP_919301.2 | |||
XM_017014362.1 | 141 | UTR 5 | XP_016869851.1 | |||
XM_017014363.1 | 141 | Intron | XP_016869852.1 | |||
XM_017014364.1 | 141 | Intron | XP_016869853.1 | |||
XM_017014365.1 | 141 | Intron | XP_016869854.1 | |||
XM_017014366.1 | 141 | Intron | XP_016869855.1 |