Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGGACTCATGGTATATTATGGAA[A/G]CAAACTTTATTTTTGCCTATCATTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612299 MIM: 611728 | ||||||||||||||||||||
Literature Links: |
COMMD9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COMMD9 - COMM domain containing 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRR5L - proline rich 5 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160167.1 | Intron | NP_001153639.1 | ||||
NM_001160168.1 | Intron | NP_001153640.1 | ||||
NM_001160169.1 | Intron | NP_001153641.1 | ||||
NM_024841.4 | Intron | NP_079117.3 |