Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGGCCGCGCCGCGTGACCGAGCT[C/G]GCGCACGCGGACCCGCCAGGGCAGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613691 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRAMD4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GRAMD4 - GRAM domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015124.4 | Intron | NP_055939.1 | ||||
XM_005261396.3 | Intron | XP_005261453.1 | ||||
XM_006724169.1 | Intron | XP_006724232.1 | ||||
XM_006724170.3 | Intron | XP_006724233.1 | ||||
XM_011530011.1 | Intron | XP_011528313.1 | ||||
XM_011530012.1 | Intron | XP_011528314.1 | ||||
XM_011530013.2 | Intron | XP_011528315.2 | ||||
XM_011530014.2 | Intron | XP_011528316.1 | ||||
XM_017028670.1 | Intron | XP_016884159.1 |
Set Membership: |
HapMap |